Skip to Content
Merck
Codeine-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Codeine-D6 solution

Codeine-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C18H15D6NO3
Molecular Weight:
305.40
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique