
Cocaine-D3 solution

Empirical Formula (Hill Notation):
C17H18NO4D3
CAS Number:
Molecular Weight:
306.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2