
Chromium

Synonym(s):
Chromium
Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Length (mm)
Element
Quality Segment
Particle Size (µm)
Showing 1-20 of 92
Page 1 of 5
Compare
Product Number
Description
Pricing
Page 1 of 5