![Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%](/deepweb/assets/sigmaaldrich/product/structures/119/817/3f1e87d4-53c8-4222-88ea-6eb679039c21/640/3f1e87d4-53c8-4222-88ea-6eb679039c21.png)
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
![Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%](/deepweb/assets/sigmaaldrich/product/structures/119/817/3f1e87d4-53c8-4222-88ea-6eb679039c21/640/3f1e87d4-53c8-4222-88ea-6eb679039c21.png)
Synonym(s):
1,5-Cyclooctadienerhodium(I) chloride dimer, Bis(1,5-cyclooctadiene)dirhodium(I) dichloride, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]dirhodium, Rhodium(I) chloride 1,5-Cyclooctadiene complex dimer, [Rh(COD)Cl]2
Empirical Formula (Hill Notation):
C16H24Cl2Rh2
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing