
Bromperidol decanoate

Synonym(s):
Decanoic acid 4-(4-bromophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl ester
Empirical Formula (Hill Notation):
C31H41BrFNO3
CAS Number:
Molecular Weight:
574.56
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2