
Benzeneruthenium(II) chloride dimer

Synonym(s):
Bis[(benzene)dichlororuthenium], Dibenzenetetrachlorodiruthenium
Linear Formula:
Ru2(C6H6)2Cl4
CAS Number:
Molecular Weight:
500.18
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing