
Azacitidine Related Compound C

Synonym(s):
1-Amidino-3-β-D-ribofuranosylurea monopicrate, 1-β-D-Ribofuranosyl-3-guanylurea picrate
Empirical Formula (Hill Notation):
C7H14N4O5 · C6H3N3O7
CAS Number:
Molecular Weight:
463.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing