
Antibodix® WCX-NP (5 μm) Weak Cation Exchange HPLC Columns

Products
Product Category
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing