Skip to Content
Merck
Aniline Blue diammonium salt certified by the BSC

Aniline Blue diammonium salt

Aniline Blue diammonium salt certified by the BSC
Empirical Formula (Hill Notation):
C37H32N5O9S3
CAS Number:
Molecular Weight:
786.87

Products

Product Category
Color
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing