
Akt Inhibitor V, Triciribine

Synonym(s):
InSolution Akt Inhibitor V, Triciribine
Empirical Formula (Hill Notation):
C13H16N6O4
CAS Number:
Molecular Weight:
320.30
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing