
8-Anilino-1-naphthalenesulfonic acid

Synonym(s):
N-Phenyl peri acid, ANS, ANSA
Linear Formula:
C6H5NHC10H6SO3H
CAS Number:
Molecular Weight:
299.34
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing