Skip to Content
BZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
6-Aminohexanoic acid
Synonym(s):
6-Aminohexanoic acid, 6-Aminocaproic acid, ε-Aminocaproic acid, EACA
Linear Formula:
H
2
N(CH
2
)
5
CO
2
H
CAS Number:
60-32-2
Molecular Weight:
131.17
EC Number:
200-469-3
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Puriss (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
8.00145
6-Aminohexanoic acid,
for synthesis
Expand
View Pricing
07260
6-Aminohexanoic acid,
≥98.5% (NT)
Expand
View Pricing