
5-Hydroxytryptophol

Synonym(s):
2-(5-Hydroxy-3-indolyl)ethyl alcohol
Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing