
4-Aminophthalhydrazide

Synonym(s):
6-Amino-2,3-dihydro-1,4-phthalazinedione, Isoluminol
Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing