
3-Fluorophenylboronic acid

Synonym(s):
(3-Fluorophenyl-1-yl)boronic acid, 3-Fluorobenzeneboronic acid, m-Fluorophenylboronic acid, m-fluoro-Benzeneboronic acid
Linear Formula:
FC6H4B(OH)2
CAS Number:
Molecular Weight:
139.92
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing