Skip to Content
Merck
3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester ≥95%, powder

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester ≥95%, powder
Synonym(s):
N-Succinimidyl 3-(2-pyridyldithio)propionate, SPDP
Empirical Formula (Hill Notation):
C12H12N2O4S2
CAS Number:
Molecular Weight:
312.36
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing