
1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene

Synonym(s):
1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene, SIMes
Empirical Formula (Hill Notation):
C21H26N2
CAS Number:
Molecular Weight:
306.44
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing