Skip to Content
Merck
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, polymer-bound 200-400 mesh, extent of labeling: 2.6 mmol/g loading, 2 % cross-linked with divinylbenzene

1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, polymer-bound

1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, polymer-bound 200-400 mesh, extent of labeling: 2.6 mmol/g loading, 2 % cross-linked with divinylbenzene
Synonym(s):
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine bound to polystyrene, 1,5,7-Triazabicyclo[4.4.0]dec-5-ene–polystyrene crosslinked with 2% DVB, ‘7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene’ on polystyrene

Products

Product Category
Chain Type
Reaction Type
Reactivity