Skip to Content
Merck
1,1′-Ferrocenedimethanol 97%

1,1′-Ferrocenedimethanol

1,1′-Ferrocenedimethanol 97%
Empirical Formula (Hill Notation):
C12H14FeO2
CAS Number:
Molecular Weight:
246.08

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing