Skip to Content
Merck
All Photos(1)

Key Documents

50695-U

Hamilton® PRP-C18 (12 - 20 µm) HPLC Columns

L × I.D. 20 mm × 4.6 mm, stainless steel, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Hamilton PRP-C18 Preparative Guard Cartridge, 12-20 μm particle size, L × I.D. 20 mm × 4.6 mm, stainless steel, pkg of 2 ea

material

stainless steel

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

20 mm × 4.6 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

C18 (octadecyl) phase

particle size

12-20 μm

pore size

100 Å

operating pH

1-14

separation technique

reversed phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service