Key Documents
Z657964
Hettich® centrifuge rotors
for UNIVERSAL 320/320R Centrifuge, Fixed Angle Rotor, 30 place
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
feature
lid
to hold 30 x 0.2/2.0 mL micolite tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1689-A
parameter
21,255 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Universal 320/320R
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG