Skip to Content
Merck
All Photos(1)

Key Documents

SML3806

Sigma-Aldrich

Cefditoren sodium

≥95% (HPLC)

Synonym(s):

(6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(1Z)-2-(4-methyl-5-thiazolyl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Cefditoren Acid Sodium Salt, ME 1206 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N6NaO5S3
CAS Number:
Molecular Weight:
528.56
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cefditoren sodium is orally available, broad spectrum third-generation cephalosporin antibiotic. Cefditoren sodium is active against both gram-negative and gram-positive bacteria. It is commonly used for treatment of chronic bronchitis, community-acquired pneumonia, pharyngitis/tonsillitis, and uncomplicated skin and skin-structure infections.
Orally available, broad spectrum third-generation cephalosporin antibiotic

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetic changes of cefdinir and cefditoren and its molecular mechanisms in acute kidney injury in rats
The Journal of Pharmacy and Pharmacology, 70(11), 1503-1512 (2018)
Enhanced in vivo activity of cefditoren in pre-immunized mice against penicillin-resistant S. pneumoniae (serotypes 6B, 19F and 23F) in a sepsis model
PLoS ONE, 5(8) (2010)
A Tamura et al.
Antimicrobial agents and chemotherapy, 32(9), 1421-1426 (1988-09-01)
ME1207 (pivaloyloxymethyl ester of ME1206) is a new oral cephalosporin. ME1206 is (6R,7R)-7-[(Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)- acetamido]-3-[(Z)-2-(4-methylthiazol-5-yl)-ethyl]-cephem-4-carboxy lic acid. The susceptibilities of about 1,600 clinical isolates to ME1206 were determined by the agar dilution method. ME1206 showed a broad spectrum of activity against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service