Skip to Content
Merck
All Photos(1)

Key Documents

Sf9 TitreHigh AC free

5011001, insect ovary, Spherical

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Sf9 TitreHigh AC free, 05011001

biological source

insect ovary

growth mode

Adherent

karyotype

Not specified

morphology

Spherical

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Spodoptera frugiperda pupal ovarian tissue, serum-free

Cell Line Description

This cell line is a derivative of the Sf9 cell line which has been adapted to TiterHigh Sf Insect Medium (Sigma I5408). TiterHigh Sf Insect Medium is an animal component-free, protein-free formulation designed specifically for the Sf9 (Spodoptera frugiperda) insect cell line. The cell line Sf9 is derived from pupal ovarian tissue of spodoptera frugiperda. The Sf9 cell line is highly susceptible to Baculovirus infection and is used in the production of protein products genetically manipulated into Baculovirus vector systems.

Application

Virus studies: Baculovirus

Culture Medium

TiterHigh Sf Insect Medium (Sigma I5408) no supplements.

Subculture Routine

Maintain cultures between 3x100,000 to 1x1,000,000 cells/ml; 27°C. Gently resuspend cells in old culture medium by dislodging the cells by pipetting, or gentle agitation, and resuspend in fresh medium. Cells should stick to the culture flask. Freeze in 7.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

TiterHigh is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service