Skip to Content
Merck
All Photos(2)

Documents

B352-05

Sigma-Aldrich

Bovine Pulmonary Artery Smooth Muscle Cells: BPASMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

bovine pulmonary artery (tunica intima and media)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 60

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

BPASMC have been used in studies relevant to pulmonary hypertension characterized by hyperproliferation of pulmonary artery smooth muscle cells, the process shown to be mediated by hypoxia-induced expression of Bcl-xl (Suzuki, 2007). Additionally, they were used to elucidate the PKC-α dependent mechanisms of proMMP-2 activation and other signaling events in smooth muscle cells under pro-inflammatory conditions related to cell growth (Chakraborti, 2009; Roy, 2011, 2013; Dey, 2012; Chowdhury, 2013; Shaikh, 2013).

Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

cell growth, signaling, receptor transactivation and suppression, vascular remodeling

Components

Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the BPASMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service