Skip to Content
Merck
All Photos(1)

Key Documents

360R-1

Sigma-Aldrich

Hemoglobin A (EPR3608) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EPR3608, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (360R-14)
vial of 0.5 mL concentrate (360R-15)
bottle of 1.0 mL predilute (360R-17)
vial of 1.0 mL concentrate (360R-16)
bottle of 7.0 mL predilute (360R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

bone marrow, placenta

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Hemoglobin alpha chain belongs to the globin family and is involved in oxygen transport from the lung to the various peripheral tissues. Hemoglobin A is comprised of two alpha chains and two beta chains, whereas hemoglobin A2 is comprised of two alpha chains and two delta chains. Immunohistochemical localization of hemoglobin is excellent as an erythroid marker for the detection of immature, dysplastic, and megaloblastic erythroid cells in myeloproliferative disorders, such as erythroleukemia. In contrast, myeloid cells, lymphoid cells, plasma cells, histiocytes, and megakaryocytes stain negative with anti-hemoglobin A. Anti-hemoglobin A, combined with antibodies against CD34, CD117, CD68, and MPO can be helpful in distinguishing between reactive extramedullary hematopoiesis and that seen in neoplastic myeloid disorders in spleen. Anti-hemoglobin A is limited to expression by erythroid precursors in bone marrow and is therefore of assistance in calculating percentages of erythroid precursors.

Quality


IVD

IVD

IVD

RUO

Linkage

HEMOGLOBIN A Positive Control Slides, Product No. 360S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis P O'Malley et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 18(12), 1550-1561 (2005-08-25)
Spleen is a common site of extramedullary hematopoiesis. Extramedullary hematopoiesis seen in non-neoplastic conditions can occasionally be extensive and raise concerns for a myeloid neoplasm. We compared the morphologic and immunohistochemical features of splenic hematopoietic proliferations seen in neoplastic myeloid
Cherie H Dunphy et al.
Applied immunohistochemistry & molecular morphology : AIMM, 15(2), 154-159 (2007-05-26)
Accurate bone marrow (BM) blast counts (BCs) are essential for diagnosis (dx) of myelodysplasia (MDS), MDS/myeloproliferative (MDS/MPD) disease, or acute myeloid leukemia (AML), and may be difficult in hemodiluted bone marrow aspirates (BMAs). Erythroid precursors (EPs) may be indistinguishable from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service