Skip to Content
Merck
All Photos(1)

Key Documents

Y0001378

Naringin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Beilstein:
102012
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

naringin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)CC(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3/t10-,16?,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

DFPMSGMNTNDNHN-JJLSSNRUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naringin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Naringin, a flavanoid in grapefruit and other citrus fruits, potently inhibits intestinal organic anion-transporting polypeptide 1A2 (OATP1A2). Grapefruit juice thereby reduces bioavailability of many pharmacological agents taken at the same time.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustafa Ozyürek et al.
Chemico-biological interactions, 212, 40-46 (2014-02-11)
Recent interest in polyphenolic antioxidants due to their involvement in health benefits has led to the investigation of new polyphenolic compounds with enhanced antioxidant activity. Naringin (4',5,7-trihydroxyflavanone-7-β-l-rhamnoglucoside-(1,2)-α-d-glucopyranoside) is one of the major flavanones in citrus and grapefruit. The present study
M A Nguyen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 68, 36-42 (2014-12-09)
Naringin is considered the major causative ingredient of the inhibition of intestinal drug uptake by grapefruit juice. Moreover, it is contained in highly dosed nutraceuticals available on the market. A controlled, open, randomized, crossover study was performed in 10 healthy
Bidya Dhar Sahu et al.
Toxicology and applied pharmacology, 277(1), 8-20 (2014-03-19)
Gentamicin-induced nephrotoxicity has been well documented, although its underlying mechanisms and preventive strategies remain to be investigated. The present study was designed to investigate the protective effect of naringin, a bioflavonoid, on gentamicin-induced nephrotoxicity and to elucidate the potential mechanism.
Eunju Leem et al.
The Journal of nutritional biochemistry, 25(7), 801-806 (2014-05-07)
This study investigated the effect of naringin, a major flavonoid in grapefruit and citrus fruits, on the degeneration of the nigrostriatal dopaminergic (DA) projection in a neurotoxin model of Parkinson's disease (PD) in vivo and the potential underlying mechanisms focusing
Feng-Quan Tan et al.
BMC plant biology, 15, 89-89 (2015-04-08)
Polyploidy has often been considered to confer plants a better adaptation to environmental stresses. Tetraploid citrus rootstocks are expected to have stronger stress tolerance than diploid. Plenty of doubled diploid citrus plants were exploited from diploid species for citrus rootstock

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service