Skip to Content
Merck
All Photos(1)

Documents

Y0000653

Tamsulosin Racemate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-[2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O5S · HCl
CAS Number:
Molecular Weight:
444.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tamsulosin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(C1=C(OC)C=CC(CC(NCCOC2=C(OCC)C=CC=C2)C)=C1)(N)=O.[H]Cl

InChI

1S/C20H28N2O5S.ClH/c1-4-26-17-7-5-6-8-18(17)27-12-11-22-15(2)13-16-9-10-19(25-3)20(14-16)28(21,23)24;/h5-10,14-15,22H,4,11-13H2,1-3H3,(H2,21,23,24);1H

InChI key

ZZIZZTHXZRDOFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tamsulosin Racemate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuo Yang et al.
The AAPS journal, 16(4), 860-871 (2014-05-31)
Multi-pellet formulations are advantageous for the controlled release of drugs over single-unit dosage forms. To understand the diffusion controlled drug release mechanism, the pellet structure and drug release from a single pellet (not at dose level) were studied using synchrotron
Thomas Kunit et al.
American journal of physiology. Renal physiology, 307(7), F823-F832 (2014-07-25)
Smooth muscle contraction may be critical for lower urinary tract symptoms (LUTS) in patients with benign prostate hyperplasia and requires stable anchorage of the cytoskeleton to the cell membrane. These connections are regulated by focal adhesion kinase (FAK). Here, we
Nannan Chu et al.
Clinical therapeutics, 37(2), 462-472 (2014-12-04)
The primary aim of this study was to evaluate whether there was clinically significant pharmacokinetic (PK) interaction between finasteride and tamsulosin in healthy Chinese male subjects. This was an open-label, randomized, 3-period, crossover study. Subjects received single and multiple doses
S Kimsakulvech et al.
Andrologia, 47(3), 354-359 (2014-04-02)
Male temporary contraception is occasionally required in some animals. Alpha1-adrenoceptor antagonist (tamsulosin) can cause ejaculation disorder. Two sets of Latin square were applied to six male goats to received either normal saline, dimethylsulphoxide or tamsulosin (179.8 nmol kg(-1) ) at 1-week interval.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service