Skip to Content
Merck
All Photos(1)

Key Documents

NIST1761A

Low alloy steel

NIST® SRM® 1761a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 1761a contains a disk that measures ~ 34 mm in diameter and 19 mm in thickness. For more information, please refer to the COA and SDS.

SRM 1761A_cert

SRM 1761A_SDS

Application

The Standard Reference Material (SRM) Low Alloy Steel is intended for use in evaluating chemical and instrumental analysis methods and calibrating instrumental analytical techniques.

Features and Benefits

  • Certified values, reference values and information value for constituents are provided.
  • Expiration details, instructions for use, handling and storage are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Boron (B), Aluminum (Al), Silicon (Si), Phosphorus (P), Sulfur (S), Titanium (Ti), Vanadium (V), Chromium (Cr), Manganese (Mn), Nickel (Ni), Copper (Cu), Zirconium (Zr), Niobium (Nb), Molybdinum (Mo)

Reference/Informational value is provided for the following analytes.
Carbon (C), Nitrogen (N), Cobalt (Co), Arsenic (As), Tin (Sn), Antimony (Sb), Tantallum (Ta), Iron (Fe)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service