Skip to Content
Merck
All Photos(1)

Documents

MABN2550

Sigma-Aldrich

Anti-GM2 ganglioside Antibody, clone GMB28

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

GMB28
monoclonal

purified by

(Ligatrap)

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgMκ

General description

Purified GM2 ganglioside attached to Salmonella minnesota.

Specificity

Clone GMB28 is a mouse monoclonal antibody that specifically detects GM2 ganglioside.

Application

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected GM2 ganglioside in human prostate cancer tissue sections.

ELISA Analysis: A representative lot detected GM2 ganglioside in ELISA applications (Kotani, M., et. al. (1992). Biochim Biophys Acta. 1117(1):97-103).

Immunohistochemistry Analysis: A representative lot detected GM2 ganglioside in Immunohistochemistry applications (Oqawa, Y., et. al. (2017). Sci Rep. 7:40518).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected GM2 ganglioside in human kidney tissue sections.

Physical form

Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human. Predicted to react with Bovine based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service