Skip to Content
Merck
All Photos(1)

Key Documents

900886

Sigma-Aldrich

SYNTHIA Retrosynthesis Software

Synonym(s):

Chemical pathway selection tool, Retrosynthesis software tool

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

Agency

ISO/IEC 27001 (Data Security)

description

Software as a Service (Saas)

application(s)

pharma/biopharma processes
pharmaceutical

compatibility

configured for Retrosynthesis

General description

SYNTHIA retrosynthesis software assists organic chemists to develop robust synthetic routes to their target molecule.

Application

SYNTHIA retrosynthesis software assists organic chemists to develop robust synthetic routes to their target molecule. Using expert-coded rules and AI-powered algorithms developed over 15 years, this decision support tool helps to reduce the risk of failure in the lab, support more sustainable chemistry, and spur innovation. In laboratory validation, SYNTHIA retrosynthesis software found robust and reliable pathways that reduce synthetic steps, increase yields, and decrease costs for both known and novel targets.

Design customized synthesis pathways starting from commercially available starting materials, building blocks, synthons, or proprietary inventory. Refine and optimize the results based on your preferences and project objectives.

Features and Benefits

  • Simplify Route Design and efficiently scan hundreds of pathways to identify the best synthesis options for your project needs.
  • Benefit from expert-coded rules and AI-powered algorithms to reduce the risk of failure, support sustainable chemistry, and spur innovation.
  • Build a Shopping List to generate a list of commercially available starting materials for your synthesis.
  • ISO 27001 Certified to safeguard and guarantee the confidentiality of your data.

Legal Information

Synthia is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service