Skip to Content
Merck
All Photos(1)

Key Documents

8.22156

Sigma-Aldrich

Tetramethylammonium chloride

for synthesis

Synonym(s):

Tetramethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4NCl
CAS Number:
Molecular Weight:
109.60
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-880-8
NACRES:
NA.22
form:
crystals
Pricing and availability is not currently available.

Quality Level

form

crystals

potency

50 mg/kg LD50, oral (Rat)
537 mg/kg LD50, skin (Rat)

pH

6-8 (20 °C, 100 g/L in H2O)

mp

268 °C

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Cl-].[N+](C)(C)(C)C

InChI

1S/C4H12N.ClH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

OKIZCWYLBDKLSU-UHFFFAOYSA-M

Application

  • Chromatography applications: Tetramethylammonium chloride serves as an effective ion-pairing agent in the separation of full and empty adeno-associated virus capsids using anion-exchange chromatography, contributing to its role in virology research (Kurth et al., 2024).

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - STOT SE 1 Oral

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service