Skip to Content
Merck
All Photos(1)

Key Documents

5.08225

Sigma-Aldrich

(Z)-4-Hydroxytamoxifen

≥99% (HPLC), liquid, estrogen receptor modulator, Calbiochem

Synonym(s):

InSolution Tamoxifen, 4-Hydroxy-, ( Z)-, ( Z)-4-(1-(4-(Dimethylaminoethoxy)phenyl)-2-phenyl-1-butenyl)phenol, ( Z)-4-(1-(4-(2-(Dimethylamino)ethoxy)phenyl)-2-phenylbut-1-enyl)phenol, (Z)-4-Hydroxytamoxifen, (Z)-4-OH-TAM, trans-4-Hydroxytamoxifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29NO2
CAS Number:
Molecular Weight:
387.51
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

(Z)-4-Hydroxytamoxifen, InSolution, ≥99%

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

N(CCOc1ccc(cc1)\C(=C(\CC)/c3ccc(cc3)O)\c2ccccc2)(C)C

InChI

1S/C26H29NO2/c1-4-25(20-10-14-23(28)15-11-20)26(21-8-6-5-7-9-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25-

InChI key

DODQJNMQWMSYGS-QPLCGJKRSA-N

General description

A 10 mM (2 mg/516 µL) sterile-filtered solution of (Z)-4-Hydroxytamoxifen in ethanol.
A cell-permeable, active metabolite of Tamoxifen (Cat. No. 579000) that acts as a potent inhibitor of PKC. It is more potent than the parent compound and inhibits PKC by modifying its catalytic domain.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Estrogen receptor

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Physical form

A 10 mM (2 mg/516 µL) sterile-filtered solution of (Z)-4-Hydroxytamoxifen in EtOH.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C. Use only anhydrous, 200 proof ethanol for further dilutions.

Other Notes

Gundimeda, U., et al. 1996. J. Biol. Chem.271, 13504.
Ye, Q. and Bodell, W.J. 1996. Carcinogenesis17, 1747.
Laser, R., et al. 1985. Eur. J. Cancer Clin. Oncol.21, 985.
Jordan, V.C., et al. 1978. J. Toxicol. Environ. Health4, 364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service