Skip to Content
Merck
All Photos(2)

Documents

1.17953

Supelco

Zinc Test Strips, colorimetric

greener alternative

0-50 mg/L (Zn), MQuant®

Synonym(s):

Zinc chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Zinc Test, colorimetric, 0-50 mg/L (Zn), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

zinc

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

0-50 mg/L (Zn)

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric with test strips, 0 - 4 - 10 - 20 - 50 mg/L (Zn), MQuant®

The MQuant® Zinc test strips can be used to measure zinc and its compounds, which are used in electroplating baths, for manufacturing zinc coatings, as a mordant for dyes, as a conservation agent, as an additive in glass, enamel, and ceramic products and finally, due to its antiseptic action, in cosmetic products.On the test strip, zinc ions react with dithizone to form a red complex. The zinc concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the tin concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® Tin test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10038-1

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service