Skip to Content
Merck
All Photos(1)

Key Documents

1.04007

Millipore

D(-)-Fructose

for biochemistry

Synonym(s):

D(-)-Fructose, Laevulose, Levulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-333-3
NACRES:
NA.79
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (HPLC)

form

solid

autoignition temp.

360 °C

pH

5-6 (20 °C, 100 g/L in H2O)

mp

100-104 °C (decomposition)

solubility

790 g/L (OECD Test Guideline 105)

density

1.59 g/cm3 at 20 °C (OECD Test Guideline 109)

bulk density

700‑800 kg/m3

storage temp.

15-25°C

SMILES string

O[C@H]([C@H](O)CO)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (HPLC): ≥ 99.0 %
Identity (IR-spectrum): conforms
Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance): -93.5 - -91.0 °
Heavy metals (as Pb): ≤ 0.001 %
Glucose (HPLC): ≤ 0.5 %
Water: ≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service