Skip to Content
Merck
All Photos(1)

Documents

03-0168-00

Millipore

SMC® Human IL-5 High Sensitivity Kit

1 kit sufficient for 96 wells, input: plasma, serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

 kit sufficient for 96 wells

species reactivity

human

manufacturer/tradename

SMC®

assay range

sensitivity: 0.04 pg/mL
(lower limit of quantification)

inter-assay cv: <20%
intra-assay cv: <15%
standard curve range: 0.01 – 50 pg/mL

technique(s)

ELISA: suitable
single molecule counting: suitable

input

plasma
serum

compatibility

for use with SMCxPRO® and Erenna<TMSYMBOL></TMSYMBOL> (validated)

shipped in

wet ice

storage temp.

2-8°C

General description

The SMC® human IL-5 High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.

Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2)The Erenna and SMCxPRO® immunoassay systems detect analytes using Single Molecule Counting (SMC®) technology.

Specificity

Non-human primate plasma: 4 of 4 individuals found to be detectable.
Canine plasma: 4 of 4 individuals found to be detectable.
Rat plasma: 4 of 4 individuals found to be detectable.
Mouse plasma: 1 of 4 individuals found to be detectable.
Although IL-5 was detectable in these species, the assay has not been validated for these species.

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service