Skip to Content
Merck
All Photos(2)

Key Documents

A-194

Supelco

Amisulpride-D5 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

4-Amino-N-[[1-(ethyl-1,1,2,2,2-d5)-2-pyrrolidinyl]methyl]-5-(ethylsulfonyl)-2-methoxybenzamide, N-[[1-(Pentadeuterioethyl)-2-pyrrolidinyl]methyl]-2-methoxy-4-amino-5-(ethylsulfonyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22D5N3O4S
CAS Number:
Molecular Weight:
374.51
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

General description

This stable-labeled Certified Spiking Solution® is suitable for quantitation of amisulpride in a variety of applications from therapeutic drug monitoring, forensic analysis and clinical toxicology to urine drug testing. Amisulpride, sold under the brand name Solian among others, is an atypical antipsychotic medication used to treat schizophrenia and dysthymia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service