610002
Avanti
PC Membranes 0.03um
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder PC Membrane, Polycarbonate Membranes 0.03μm 19mm (100/pk)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
PC Membranes are the filter membranes with smooth and flat surfaces with track-etched uniform cylindrical pores.
Polycarbonate Membrane for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Application
PC Membranes 0.03um has been used to extrude lipid vesicles.
Biochem/physiol Actions
PC Membrane exhibits fine mechanical properties, chemical stability, and non-auto fluorescent properties. It is extensively used for the identification of various bacteria in immunofluorescent direct count method.
Components
Whatman® Nuclepore polycarbonate hydrophilic membrane 800307
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva
also commonly purchased with this product
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mechanism of four de novo designed antimicrobial peptides
Test, 291(49), 25706- 25715 (2016)
Highly sensitive detection of Salmonella typhi using surface aminated polycarbonate membrane enhanced-ELISA
Biosensors And Bioelectronics, 31(1), 37-43 (2012)
Response of MG63 osteoblast-like cells onto polycarbonate membrane surfaces with different micropore sizes
Biomaterials, 25(19), 4699-4707 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service