Z112178
32mm Round cell window
KBr, Drilled
Synonym(s):
32mm Diameter Round Optical Cell Window
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Recommended Products
material
KBr
packaging
pkg of (contains 1 window)
technique(s)
IR spectroscopy: suitable
diam. × thickness
32 mm × 3 mm
Looking for similar products? Visit Product Comparison Guide
General description
Cut and polished for demountable liquid and gas cell holders
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service