Skip to Content
Merck
All Photos(2)

Key Documents

GF57214087

Bismuth

lump, 7 mm max. lump size, weight 50 g, purity 99.999%

Synonym(s):

BI006110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.999%

form

lumps

manufacturer/tradename

Goodfellow 572-140-87

resistivity

129 μΩ-cm, 20°C

weight

50 g

max. lump size

7 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service