Skip to Content
Merck
All Photos(1)

Key Documents

925411

Sigma-Aldrich

3-Mercapto-1-hexanol

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14OS
CAS Number:
Molecular Weight:
134.24
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

storage temp.

2-8°C

SMILES string

CCCC(S)CCO

InChI

1S/C6H14OS/c1-2-3-6(8)4-5-7/h6-8H,2-5H2,1H3

InChI key

TYZFMFVWHZKYSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service