912190
Hydroxyapatite granules
0.5-1.0 mm grain size
Synonym(s):
Apatite hydroxide, Ca10(PO4)6(OH)2, Hydroxyapatite, Hydroxylapatite, Pentacalcium hydroxide triphosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Ca10(PO4)6(OH)2
UNSPSC Code:
12352305
NACRES:
NA.23
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Handling:
Avoid contact with eyes and skin. Avoid formation of dust and aerosols. Provide appropriate exhaust ventilation at place where dust is formed.
Storage:
Keep store in clean, dry place. Keep work area free of spills. Keep the container tightly closed. Avoid freezing
Avoid contact with eyes and skin. Avoid formation of dust and aerosols. Provide appropriate exhaust ventilation at place where dust is formed.
Storage:
Keep store in clean, dry place. Keep work area free of spills. Keep the container tightly closed. Avoid freezing
Hydroxyapatite and tricalcium phosphate are bioactive ceramic materials and they find applications as bone grafts, fillers and coating material for metal implants.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hydroxyapatite/graphene-nanosheet composite coatings deposited by vacuum cold spraying for biomedical applications: Inherited nanostructures and enhanced properties.
Li Y, et al.
Carbon, 67, 250-259 (2014)
Sahar Vahabzadeh et al.
Acta biomaterialia, 17, 47-55 (2015-02-02)
In this work we have investigated the effects of strontium (Sr) dopant on in vitro protein release kinetics and in vivo osteogenic properties of plasma sprayed hydroxyapatite (HA) coatings, along with their dissolution behavior. Plasma sprayed HA coatings are widely
Sophie C Cox et al.
Materials science & engineering. C, Materials for biological applications, 47, 237-247 (2014-12-11)
A systematic characterisation of bone tissue scaffolds fabricated via 3D printing from hydroxyapatite (HA) and poly(vinyl)alcohol (PVOH) composite powders is presented. Flowability of HA:PVOH precursor materials was observed to affect mechanical stability, microstructure and porosity of 3D printed scaffolds. Anisotropic
Lu Xie et al.
Materials science & engineering. C, Materials for biological applications, 59, 1007-1015 (2015-12-15)
The ideal bone tissue engineering scaffolds are long-cherished with the properties of interconnected macroporous structures, adjustable degradation and excellent biocompatibility. Here, a series of porous α/β-tricalcium phosphate (α/β-TCP) biphasic bioceramics with different phase ratios of α-TCP and β-TCP were successfully
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service