Antibody Generation: Monoclonal vs Polyclonal Antibodies
Antibodies are normally produced by B cells, which are part of the immune system, in response to the introduction of foreign substances, such as infectious agents, into the animal’s body. The antibodies bind to the antigens that cause their generation and flag them for destruction, thus helping to fight infection. This inherent ability of the animal’s body can be leveraged to generate antibodies that bind to specific molecules.
Target-specific antibodies can be used to isolate and identify molecules of interest. Antibodies have become one of the most important tools in life science research, allowing the detection, quantitation, and determination of changes in proteins and other molecules with respect to time and other perturbations.
Read on to learn differentiations between monoclonal vs polyclonal antibodies including the differences in the way these antibodies are generated.
Monoclonal vs Polyclonal Antibodies
There are a variety of differences between monoclonal vs polyclonal antibodies, such as how they are generated. Many of the antibodies used in immunochemical techniques are raised by repeated immunization of a suitable animal, e.g., rabbit, goat, donkey, or sheep, with an appropriate antigen. Serum is harvested at the peak of antibody production. Specific IgG concentrations of approximately 1 to 10 mg/mL serum can be obtained by this method. Weakly antigenic molecules may require the addition of an adjuvant, which allows for the slow release of the antigen, making it more readily trapped by macrophages. Smaller molecules, such as drugs, must be coupled to more antigenic structures (i.e. carrier proteins) to stimulate an immune response.
One characteristic of large antigen molecules is that they induce the activation of many antibody-producing B cell clones in the immunized animal. This polyclonal mixture of resulting antibodies may then recognize a variety of epitopes on the antigen, which can be a useful feature in some experimental procedures. Because these polyclonal mixtures of antibodies react with multiple epitopes on the surface of the antigen, they will be more tolerant of minor changes in the antigen, e.g., polymorphism, heterogeneity of glycosylation, or slight denaturation, than will monoclonal (homogenous) antibodies. Depending upon the antigen that is used to create the antibody, one may use polyclonal antibodies to identify proteins of high homology to the immunogen protein or to screen for the target protein in tissue samples from species other than that of the immunogen. Along the same lines, it is especially important when working with polyclonal antibodies to learn as much as possible about the immunogen that has been used for production of the polyclonal antibody and the potential for undesired cross-reactivity within the sample being analyzed. Peptide immunogens are often used to generate polyclonal antibodies that target unique epitopes, especially for protein families of high homology.
Figure 1. Monoclonal vs Polyclonal Antibodies GenerationDiagram representing production of monoclonal vs polyclonal antibodies. Starting point is a large antigen immunogen that yields multiple epitope polyclonal antibodies. Arrows go in 2 paths, one goes to a small peptide immunogen with fewer, restricted epitope polyclonal antibodies while the other goes to a point that isolates and fuses B cells to the hybridoma line and screening. Another arrow goes from the small peptide immunogen to the hybridoma. From there, an arrow goes to another point representing a monoclonal antibody with one epitope. Off to the side there is a cell with an antibody on it representing that each B cell only produces antibodies to one epitope.
A homogeneous population of antibodies (i.e. monoclonal antibodies) can be raised by fusion of B lymphocytes with immortal cell cultures to produce hybridomas. Hybridomas will produce many copies of the exact same antibody. This impressive phenomenon has been instrumental in the development of antibodies for diagnostic applications because monoclonal antibodies react with one epitope on the antigen. However, they are more vulnerable to the loss of epitope through chemical treatment of the antigen than are polyclonal antibodies. This can be offset by pooling two or more monoclonal antibodies to the same antigen.
Polyclonal Antibodies Properties
- Polyclonal antibodies often recognize multiple epitopes, making them more tolerant of small changes in the nature of the antigen. Polyclonal antibodies are often the preferred choice for detection of denatured proteins.
- Polyclonal antibodies may be generated in a variety of species, including rabbit, goat, sheep, donkey,
chicken, and others, giving the users many options in experimental design. - Polyclonal antibodies are sometimes used when the nature of the antigen in an untested species is not known.
- Polyclonal antibodies target multiple epitopes and so they generally provide more robust detection.
Monoclonal Antibodies Properties
- Because of their specificity, monoclonal antibodies are excellent as the primary antibody in an assay, or for detecting antigens in tissue, and will often result in significantly less background signal than polyclonal antibodies.
- When compared to that of polyclonal antibodies, homogeneity of monoclonal antibodies is very high.
- If experimental conditions are kept constant, results from monoclonal antibodies will be highly reproducible between experiments.
- Specificity of monoclonal antibodies makes them extremely efficient for binding of antigen within a mixture of related molecules, such as in the case of affinity purification.
Advantages and Disadvantages of Monoclonal vs Polyclonal Antibodies
Advantages | Disadvantages | |
---|---|---|
Polyclonal Antibodies |
|
|
Monoclonal Antibodies |
|
|
Clone Numbers
Each clone number represents a specific cell line that was used to produce the antibody. Since antibodies are produced by more than one host, each cloned cell line receives a unique clone number. Each hybridoma cell clone produces only one single pure antibody type.
- An animal injected with an antigen will generate multiple antibodies to many epitopes. Since antibodies are produced by B cells, a single clone of B cells can produce antibodies to only a single epitope.
- Monoclonal antibodies are derived from a single clone of cells and can be generated in larger quantities.
- Polyclonal antibodies contain multiple clones of antibodies produced to different epitopes on the antigen. For example, if there are four epitopes on the antigen then four different clones of antibodies will be produced.
- Different antibody clones may have different properties and may even be of different isotypes. They may also work in different applications. Hence, it is best to select an antibody clone that will work optimally in your choice of application.
- It is important to recognize that a clone number is not synonymous with the lot number, which often indicates the date of manufacture.
Antibody Formats
As the name implies, the antibody format refers to the presentation or purification state of the antibody. Various formats are described below:
Polyclonal Antibodies
Polyclonal antibodies are often available in relatively unpurified formats, and are referred to as “antiserum” or simply as “serum”. Antiserum refers to the blood from an immunized host from which the clotting proteins and RBCs have been removed. The antiserum, as its name suggests, still possesses antibodies/immunoglobulins of all classes as well as other serum proteins. In addition to antibodies that recognize the target antigen, the antiserum also contains antibodies to various other antigens that can sometimes react nonspecifically in immunological assays. For this reason, raw antiserum is often subjected to purification steps, to eliminate serum proteins and to enrich the fraction of immunoglobulin that specifically reacts with the target antigen.
Antiserum is commonly purified by one of two methods: Protein A/G purification or antigen affinity chromatography.
Protein A/G purification takes advantage of the high affinity of Staphylococcus aureus protein A or Streptococcus protein G for the immunoglobulin Fc domain. While protein A/G purification eliminates the bulk of the serum proteins from the raw antiserum, it does not eliminate the nonspecific immunoglobulin fraction. As a result, the protein A/G purified antiserum may still possess undesirable cross reactivity.
Antigen affinity purification takes advantage of the affinity of the specific immunoglobulin fraction for the immunizing antigen against which it was generated. This method may be used to remove unwanted antibodies from a preparation. The preparation of antibodies is passed through a column matrix containing antigens against which the unwanted antibodies are directed. The unwanted antibodies remain bound to the column, and the effluent contains the desired, affinity-purified antibodies. Alternatively, a column matrix coupled to the desired antigen can be used. In this case, antibody directed against the coupled antigen remains bound to the column and may be then eluted using a solution that disrupts antigen-antibody binding. Unlike protein A/G purification, antigen affinity purification results in the elimination of the bulk of the nonspecific immunoglobulin fraction, while enriching the fraction of immunoglobulin that specifically reacts with the target antigen. The resulting affinity purified immunoglobulin will contain primarily the immunoglobulin of desired specificity.
Typically, affinity purified antibodies exhibit lower backgrounds than unabsorbed antibodies and this purification process is particularly important for difficult, or state-dependent epitopes. When developing polyclonal antibodies that recognize targets with post-translational modifications, the use of modification specific antigen affinity columns during the purification process can significantly improve the specificity of the antibody for state-dependent target. Depleting unmodified target protein from the serum before affinity purification (using immobilized, modified target protein) increases the specificity for the modified target. Specificity testing can then be performed to confirm that the antibody only recognizes the post-translationally modified form of the Protein.
Monoclonal Antibodies
Monoclonal antibodies may be grown in cell cultures and collected as hybridoma supernatants, or grown in mice or rats and collected as relatively unpurified ascites fluid. These can be purified through the use of protein A/G or specific antigen affinity chromatography as with polyclonal antibodies.
Unpurified antibody preparations vary significantly in specific antibody concentration. If the specific antibody concentration of a given unpurified antibody preparation is unknown, one may refer to the following “typical ranges” as a guideline for estimation:
- Polyclonal Antiserum: Specific antibody concentrations will typically range from 1–3 mg/mL.
- Hybridoma Supernatant: Specific antibody concentrations will typically range from 0.1–10.0 mg/mL.
- Ascites Fluid (unpurified): Specific antibody concentrations will typically range from 2–10 mg/mL.
Learn about another antibody format, recombinant antibodies, that is highly reproducible in our technical article on ZooMAb® recombinant monoclonal antibodies.
Biological Effects of Antibodies
Antibodies are widely used for protection from infectious agents. Most vaccines (microbial antigens) induce the production of antibodies that block infection or interfere with microbial invasion of the bloodstream. To achieve this, antibodies must be functional in the sense that they are capable of neutralization or opsonophagocytosis.
The membrane attack complex (MAC) cytolysis MAC is formed on the surface of pathogenic bacterial cell as a result of the activation of the complement system (both alternative and the classical pathways). The MAC forms transmembrane channels in bacterial walls, disrupting their phospholipid bilayer and leading to cell lysis and death.
Neutralization of Viruses
Antibodies can interfere with virion binding to receptors and block their uptake into cells. Many enveloped viruses are lysed when antiviral antibodies and the complement system disrupt membranes. Certain antibodies can also aggregate virus particles. Non-neutralizing antibodies are also produced following any viral infection. Although these antibodies bind specifically to virus particles, they do not neutralize them. On the contrary, they may enhance infectivity because the virus-antibody complex enters the cell by endocytosis. This can lead to viral replication. The type of antibody produced can influence the outcome of viral infection. For example, poliovirus can elicit IgM and IgG responses in the blood, but mucosal IgA is vital for blocking infection. The IgA neutralizes poliovirus in the intestine, the site of primary infection. Hence, the live attenuated Sabin poliovirus vaccine is more effective because it elicits a strong mucosal IgA response.
Immobilization
An antibody can be directed against cilia or flagella of motile bacteria or protozoa that results in cessation of their motility and blocks their ability to move around and spread infection.
Cytolysis
Certain antibodies can cause disruption of the microbial membrane that result in death of bacterial cells. This requires the participation of the complement system.
Opsonization
In this process, the pathogenic organism is targeted for digestion by phagocytes. The antibody binds to a receptor on the cell membrane of the bacterium, attracting phagocytes to the site. The F(ab) portion of the antibody binds to the antigen, while the Fc portion of the antibody binds to an Fc receptor on the phagocyte, facilitating phagocytosis. This process is further enhanced by the complement system.
Neutralization of Exotoxins
Antitoxin antibodies can be generated against microbial toxins. The F(ab) region of the antibody made against epitope of the binding site of an exotoxin can block the exotoxin from binding to the exotoxin receptor on the host cell membrane. This blocks the entry of the toxin into the cell.
Preventing Bacterial Adhesion to Host Cells
The body’s innate defenses can physically remove bacteria by constant shedding of surface epithelial cells from the skin and mucous membranes. However, bacteria may resist this by producing pili, cell wall adhesin proteins, and biofilm-producing capsules. The F(ab) region of the antibody can bind to the adhesive tip of the pili, the cell wall adhesins, or the capsular molecules, and blocks bacterial adhesion to host cells.
Agglutination of Microorganisms
The F(ab) sites of IgM and IgA antibodies can link microorganisms together and cause them to agglutinate. The agglutinated microorganisms can be phagocytosed more effectively.