HPLC Analysis of Hydroxychloroquine Enantiomers on Astec® CHIROBIOTIC® V
![HPLC Analysis of Hydroxychloroquine Enantiomers on Astec® CHIROBIOTIC® V application for HPLC](/deepweb/assets/sigmaaldrich/product/images/299/544/5ab06650-2477-4a8f-8399-9731b239f955/800/5ab06650-2477-4a8f-8399-9731b239f955.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (98:1:1, A:B:C)
flow rate
2 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC