Merck
GC Analysis of Resveratrol in Red Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber
GC Analysis of Resveratrol in Red Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with manual holder, needle size 24 ga
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with autosampler, needle size 24 ga
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with autosampler, needle size 23 ga
accessory
Product No.
Description
Pricing
Vials, screw top with phenolic open-top cap, pre-assembled
volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, tan PTFE/silicone septum
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with Manual Sampling, pk of 1
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 1 ea
standard
Product No.
Description
Pricing
Resveratrol
≥99% (HPLC)
CONDITIONS
sample/matrix
3 mL of red wine (California merlot) diluted 3:1 in 12% ethanol:water
SPME fiber
85 μm polyacrylate FS 23 gauge auto (57294-U)
SPME fiber
85 μm polyacrylate FS 24gauge auto (57305)
SPME fiber
85 μm polyacrylate FS 24gauge manual (57304)
extraction
immersion for 15 min at room temperature while stirring at 400 rpm, fiber gently blotted with a Kimwipe to remove excess water, (then derivatization by immersion in a 4 mL vial containing 5 μL of Sylon-BFT (fresh vial for each analysis than was allowed to equilabrate 60-90 minutes prior to use) for 20 min)
desorption process
280 °C for 2 min
column
SLB-5ms; 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
100 °C (1 min), 10 °C/min to 325 °C (3 min)
MSD interface
325 °C
scan range
m/z 40-450
carrier gas
helium, 1 mL/min, constant
liner
0.75 mm I.D., SPME
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany