Merck
  • Utility of porous graphitic carbon stationary phase in quantitative liquid chromatography/tandem mass spectrometry bioanalysis: quantitation of diastereomers in plasma.
Utility of porous graphitic carbon stationary phase in quantitative liquid chromatography/tandem mass spectrometry bioanalysis: quantitation of diastereomers in plasma.
Rapid communications in mass spectrometry : RCM (2006-05-18)
Yuan-Qing Xia, Mohammed Jemal, Naiyu Zheng, Xiaohang Shen
ABSTRACT
A major challenge in selecting an appropriate stationary phase for diastereomeric separation is that it is difficult to predict which of the commercially available stationary phases could achieve the required liquid chromatographic (LC) separation. This work describes the selection and evaluation of a porous graphitic carbon (PGC) column coupled with tandem mass spectrometry (MS/MS) for the simultaneous quantitation of an experimental drug candidate (I), its two diastereomeric metabolites (II and III), and its demethylated metabolite (IV) in rat plasma. In addition, we investigated the PGC column for the separation of another drug candidate (VI), its two diastereomeric metabolites (VII and VIII) and its ketone metabolite (IX). The PGC column showed excellent chromatographic resolution for the two diastereomers II and III, as well as for VII and VIII. In contrast, the required resolution for the diastereomers II and III could not be achieved using silica-bonded C(18), C(30), phenyl, perfluorinated, polar embedded and polar end-capped phases. The PGC column showed ruggedness with excellent reproducibility of retention times, peak symmetry and response over a period of more than 400 injections of a plasma acetonitrile-precipitation extract. Excellent accuracy and precision were achieved, with accuracy of 94-108% and intra- and inter-run precision within 9%. This work indicates that PGC is a valuable addition to the repertoire of LC columns used for quantitative LC/MS/MS bioanalysis, especially where the separation and quantitation of diastereomeric analytes is involved.
Product Description
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm, pkg of 2 ea
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 3 mm
Pricing and availability is not currently available.
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Pricing and availability is not currently available.