Merck
  • Pharmacological blockade of ASCT2-dependent glutamine transport leads to antitumor efficacy in preclinical models.
Pharmacological blockade of ASCT2-dependent glutamine transport leads to antitumor efficacy in preclinical models.
Nature medicine (2018-01-16)
Michael L Schulte, Allie Fu, Ping Zhao, Jun Li, Ling Geng, Shannon T Smith, Jumpei Kondo, Robert J Coffey, Marc O Johnson, Jeffrey C Rathmell, Joe T Sharick, Melissa C Skala, Jarrod A Smith, Jordan Berlin, M Kay Washington, Michael L Nickels, H Charles Manning
ABSTRACT
The unique metabolic demands of cancer cells underscore potentially fruitful opportunities for drug discovery in the era of precision medicine. However, therapeutic targeting of cancer metabolism has led to surprisingly few new drugs to date. The neutral amino acid glutamine serves as a key intermediate in numerous metabolic processes leveraged by cancer cells, including biosynthesis, cell signaling, and oxidative protection. Herein we report the preclinical development of V-9302, a competitive small molecule antagonist of transmembrane glutamine flux that selectively and potently targets the amino acid transporter ASCT2. Pharmacological blockade of ASCT2 with V-9302 resulted in attenuated cancer cell growth and proliferation, increased cell death, and increased oxidative stress, which collectively contributed to antitumor responses in vitro and in vivo. This is the first study, to our knowledge, to demonstrate the utility of a pharmacological inhibitor of glutamine transport in oncology, representing a new class of targeted therapy and laying a framework for paradigm-shifting therapies targeting cancer cell metabolism.
Sigma-Aldrich
Anti-Neutral amino acid transporter (ASCT2) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Slc1a7
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes