- P311 Deficiency Leads to Attenuated Angiogenesis in Cutaneous Wound Healing.
P311 Deficiency Leads to Attenuated Angiogenesis in Cutaneous Wound Healing.
P311 was identified to markedly promote cutaneous wound healing by our group. Angiogenesis plays a key role in wound healing. In this study, we sought to define the role of P311 in skin wound angiogenesis. It was noted that P311 was expressed in endothelial cells in the dermis of murine and human skin wounds. The expression of P311 was confirmed in cultured murine dermal microvascular endothelial cells (mDMECs). Moreover, it was found that knockout of P311 could attenuate the formation of tubes and motility of mDMECs significantly