- Cyclic and linear siloxanes in indoor air from several northern cities in Vietnam: Levels, spatial distribution and human exposure.
Cyclic and linear siloxanes in indoor air from several northern cities in Vietnam: Levels, spatial distribution and human exposure.
Earlier studies have reported the occurrence of cyclic and linear siloxanes in personal care and household products. Nevertheless, there is a lack of information on the occurrence of siloxanes in indoor air. In this study, four cyclic and six linear siloxanes were measured in 97 indoor air samples collected from various micro-environments in four cities in northern, Vietnam, during September 2016 to January 2017. The total concentrations of siloxanes (TSi) in particulate and gas phases ranged from 141 to 7220 μg g