- Different breeds, different blood: Cytometric analysis of whole blood cellular composition in chicken breeds.
Different breeds, different blood: Cytometric analysis of whole blood cellular composition in chicken breeds.
While haematological variation is well known in birds, variation in avian breeds (distinct morphotypes of the same species) remains unexplored. Poultry breeds, in particular, may show interesting evolutionary patterns and economically-relevant physiological differences. We performed a comparative examination of blood cellular composition in five chicken (Gallus gallus domesticus) breeds: Araucana, Booted bantam, Czech, Minorca and Rosecomb bantam. In standard-environment-reared hens whole-blood flow cytometry revealed remarkable differences in most erythrocyte- and leukocyte-related parameters. We identified two extremes: Czech, a European breed, with a low heterophil/lymphocyte (H/L) ratio and high CD4