Merck
  • Species-specific antagonism of host ISGylation by the influenza B virus NS1 protein.
Species-specific antagonism of host ISGylation by the influenza B virus NS1 protein.
Journal of virology (2010-03-12)
Gijs A Versteeg, Benjamin G Hale, Sander van Boheemen, Thorsten Wolff, Deborah J Lenschow, Adolfo García-Sastre
ABSTRACT
Interferon-stimulated expression and conjugation of the ubiquitin-like modifier ISG15 restricts replication of several viruses. Here, we established complete E1-activating, E2-conjugating, and E3 ligase-dependent expression systems for assaying both human and mouse ISGylation. We confirm that human HerC5, but not human HerC6, has ISG15 E3 ligase activity and identify mouse HerC6 as a bona fide ISG15 E3 ligase. Furthermore, we demonstrate that influenza B virus NS1 protein potently antagonizes human but not mouse ISGylation, a property dependent on B/NS1 binding the N-terminal domain of human but not mouse ISG15. Using chimeric human/mouse ISG15 constructs, we show that the B/NS1:ISG15 interaction is both necessary and sufficient to inhibit ISGylation regardless of the ligation machinery used. Inability to block ISGylation in certain species may contribute to limiting influenza B virus host range.